siRNA Details
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113323.1 | NP_001106794.1 | 20 | 2180 | ||
| GenBank mRNA | AK014852.1 | - | 20 | 2013 | ||
| BC132200.1 | - | 20 | 1973 | 40130477 | ||
| BC132202.1 | - | 20 | 1973 | 40130479 | ||

Glb1l3
Genomic Map