siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027597.4 | NP_081873.2 | 5 | 891 | ||
| NM_029069.1 | NP_083345.1 | 5 | 896 | |||
| GenBank mRNA | AB473726.1 | - | 3 | 571 | ||
| AK014851.1 | - | 5 | 897 | |||
| AK015802.1 | - | 5 | 743 | |||
| BC051149.1 | - | 5 | 751 | 6707726 | ||

Fam71d
Genomic Map