siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027597.4 | NP_081873.2 | 5, 6 | 1265 | ||
| NM_029069.1 | NP_083345.1 | 5, 6 | 1270 | |||
| GenBank mRNA | AB473726.1 | - | 3, 4 | 945 | ||
| AK014851.1 | - | 5, 6 | 1271 | |||
| AK015802.1 | - | 5, 6 | 1117 | |||
| BC051149.1 | - | 5, 6 | 1125 | 6707726 | ||

Fam71d
Genomic Map