siRNA Details
| Gene Symbol: Speer4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027609.2 | NP_081885.1 | 2 | 265 | ||
| GenBank mRNA | AK015022.1 | - | 2 | 268 | ||
| BC116901.1 | - | 2 | 291 | 40126220 | ||
| BC116903.1 | - | 2 | 291 | 40126222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer4f| Gene Symbol: Speer4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027609.2 | NP_081885.1 | 2 | 265 | ||
| GenBank mRNA | AK015022.1 | - | 2 | 268 | ||
| BC116901.1 | - | 2 | 291 | 40126220 | ||
| BC116903.1 | - | 2 | 291 | 40126222 | ||