siRNA Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199210.1 | NP_001186139.1 | 4 | 976 | ||
| NM_027627.2 | NP_081903.1 | 4 | 976 | |||
| GenBank mRNA | AF358257.1 | - | 4 | 703 | ||
| AK016443.1 | - | 6 | 972 | |||
| AK037486.1 | - | 3 | 831 | |||
| BC100350.1 | - | 4 | 722 | 30522196 | ||
| BC132305.1 | - | 4 | 976 | 40130582 | ||
| BC144751.1 | - | 3 | 778 | |||
| BC157969.1 | - | 2 | 526 | 9007291 | ||

Eva1c
Genomic Map