siRNA Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199210.1 | NP_001186139.1 | 6 | 1231 | ||
| NM_027627.2 | NP_081903.1 | 5 | 1087 | |||
| GenBank mRNA | AF358257.1 | - | 6 | 958 | ||
| AK016443.1 | - | 8 | 1227 | |||
| AK037486.1 | - | 4 | 942 | |||
| BC100350.1 | - | 5 | 833 | 30522196 | ||
| BC132305.1 | - | 6 | 1231 | 40130582 | ||
| BC144751.1 | - | 5 | 1033 | |||
| BC157969.1 | - | 4 | 781 | 9007291 | ||

Eva1c
Genomic Map