siRNA Details
Gene Symbol: 4931423N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027635.1 | NP_081911.1 | 7, 8 | 1234 | ||
GenBank mRNA | AK016475.1 | - | 7, 8 | 1235 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4931423N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027635.1 | NP_081911.1 | 7, 8 | 1234 | ||
GenBank mRNA | AK016475.1 | - | 7, 8 | 1235 |