siRNA Details
| Gene Symbol: Prss41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027644.1 | NP_081920.1 | 4, 5 | 314 | ||
| GenBank mRNA | AB049453.1 | - | 4, 5 | 314 | ||
| AK016509.1 | - | 4, 5 | 292 | |||
| AK049297.1 | - | 4, 5 | 331 | |||
| BC145586.1 | - | 4, 5 | 330 | |||
| BC150797.1 | - | 4, 5 | 330 | 9087708 | ||

Prss41
Genomic Map