siRNA Details
| Gene Symbol: Spats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027649.3 | NP_081925.2 | 3 | 366 | ||
| GenBank mRNA | AK005865.1 | - | 3 | 367 | ||
| AK016586.1 | - | 3 | 293 | |||
| AK078278.1 | - | 288 | ||||
| AK165435.1 | - | 2 | 154 | |||
| AY336501.1 | - | 3 | 254 | |||
| BC131909.1 | - | 3 | 326 | 40130186 | ||

Spats1
Genomic Map