siRNA Details
| Gene Symbol: Spats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027649.3 | NP_081925.2 | 3 | 412 | ||
| GenBank mRNA | AK005865.1 | - | 3 | 413 | ||
| AK016586.1 | - | 3 | 339 | |||
| AK078278.1 | - | 334 | ||||
| AK165435.1 | - | 2 | 200 | |||
| AY336501.1 | - | 3 | 300 | |||

Spats1
Genomic Map