siRNA Details
| Gene Symbol: Spats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027649.3 | NP_081925.2 | 4, 5 | 603 | ||
| GenBank mRNA | AK005865.1 | - | 4, 5 | 604 | ||
| AK016586.1 | - | 4, 5 | 530 | |||
| AK078278.1 | - | 525 | ||||
| AY336501.1 | - | 4, 5 | 491 | |||
| BC131909.1 | - | 4, 5 | 563 | 40130186 | ||

Spats1
Genomic Map