siRNA Details
| Gene Symbol: Speer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027650.3 | NP_081926.2 | 3 | 433 | ||
| GenBank mRNA | AK016676.1 | - | 3 | 420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer3| Gene Symbol: Speer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027650.3 | NP_081926.2 | 3 | 433 | ||
| GenBank mRNA | AK016676.1 | - | 3 | 420 | ||