siRNA Details
| Gene Symbol: Pcgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027654.2 | NP_081930.1 | 4 | 605 | ||
| GenBank mRNA | AB047007.1 | - | 4 | 655 | ||
| AK016709.1 | - | 4 | 627 | |||
| AK132448.1 | - | 4 | 651 | |||
| AK155067.1 | - | 4 | 666 | |||
| BC016195.1 | - | 4 | 605 | 4488614 | ||
| BC089460.1 | - | 4 | 644 | 30632537 | ||

Pcgf6
Genomic Map