siRNA Details
| Gene Symbol: Dmrtc1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142691.1 | NP_001136163.1 | 3 | 482 | ||
| GenBank mRNA | AK016891.1 | - | 3 | 483 | ||
| BC116811.1 | - | 2 | 116 | 40126130 | ||
| BC116813.1 | - | 2 | 116 | 40126132 | ||
| Gene Symbol: Dmrtc1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142690.1 | NP_001136162.1 | 3 | 482 | ||

Dmrtc1c1
Genomic Map