siRNA Details
| Gene Symbol: Sept12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027669.3 | NP_081945.1 | 7 | 830 | ||
| NM_029374.1 | NP_083650.1 | 7 | 830 | |||
| GenBank mRNA | AK006458.1 | - | 7 | 768 | ||
| AK016798.1 | - | 7 | 758 | |||
| AK077091.1 | - | 7 | 831 | |||
| BC115618.1 | - | 7 | 741 | 40102763 | ||
| BC115619.1 | - | 6 | 876 | 40102765 | ||

Sept12
Genomic Map