siRNA Details
| Gene Symbol: 4933413J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016807.1 | - | 6 | 632 | ||
| AK087762.1 | - | 4 | 383 | |||
| DV663125.1 | - | 549 | ||||
| HM015003.1 | - | 5 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933413J09Rik| Gene Symbol: 4933413J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016807.1 | - | 6 | 632 | ||
| AK087762.1 | - | 4 | 383 | |||
| DV663125.1 | - | 549 | ||||
| HM015003.1 | - | 5 | 603 | |||