siRNA Details
| Gene Symbol: Sh2d6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016886.1 | - | 4, 5 | 376 | ||
| BC100525.1 | - | 4, 5 | 333 | 30920200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d6| Gene Symbol: Sh2d6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016886.1 | - | 4, 5 | 376 | ||
| BC100525.1 | - | 4, 5 | 333 | 30920200 | ||