siRNA Details
| Gene Symbol: Zfp689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175163.3 | NP_780372.2 | 3 | 958 | ||
| GenBank mRNA | AK028417.1 | - | 3 | 921 | ||
| AK051667.1 | - | 3 | 772 | |||
| BC052084.1 | - | 3 | 958 | 6404620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp689| Gene Symbol: Zfp689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175163.3 | NP_780372.2 | 3 | 958 | ||
| GenBank mRNA | AK028417.1 | - | 3 | 921 | ||
| AK051667.1 | - | 3 | 772 | |||
| BC052084.1 | - | 3 | 958 | 6404620 | ||