siRNA Details
Gene Symbol: 4933407L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016730.1 | - | 3 | 707 | ||
AK037440.1 | - | 1 | 2432 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933407L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016730.1 | - | 3 | 707 | ||
AK037440.1 | - | 1 | 2432 |