siRNA Details
| Gene Symbol: Scara5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168318.1 | NP_001161790.1 | 6 | 1435 | ||
| NM_028903.2 | NP_083179.2 | 6 | 1435 | |||
| GenBank mRNA | AK016543.1 | - | 6 | 1470 | ||
| AK016912.1 | - | 6 | 1402 | |||
| AK033154.1 | - | 6 | 1402 | |||
| AK133176.1 | - | 6 | 1436 | |||
| BC016096.1 | - | 6 | 1411 | 4506472 | ||
| BC023907.1 | - | 6 | 1351 | 5321978 | ||
| BC026758.1 | - | 3 | 461 | |||
| BC032159.1 | - | 6 | 1391 | 5354657 | ||
| DQ122126.1 | - | 5 | 1085 | |||

Scara5
Genomic Map