siRNA Details
| Gene Symbol: Golga7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141983.1 | NP_001135455.1 | 6 | 746 | ||
| NM_027694.2 | NP_081970.1 | 4 | 737 | |||
| GenBank mRNA | AK016850.1 | - | 6 | 751 | ||
| AK139354.1 | - | 4 | 738 | |||
| BC049116.1 | - | 4 | 693 | 6811056 | ||

Golga7b
Genomic Map