siRNA Details
| Gene Symbol: Golga7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141983.1 | NP_001135455.1 | 6, 7 | 833 | ||
| NM_027694.2 | NP_081970.1 | 4, 5 | 824 | |||
| GenBank mRNA | AK016850.1 | - | 6, 7 | 838 | ||
| AK139354.1 | - | 4, 5 | 825 | |||
| BC049116.1 | - | 4, 5 | 780 | 6811056 | ||

Golga7b
Genomic Map