siRNA Details
| Gene Symbol: Lrrc72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177877.1 | NP_001171348.1 | 7 | 743 | ||
| NM_027699.2 | NP_081975.1 | 3 | 240 | |||
| GenBank mRNA | AK007140.1 | - | 3 | 241 | ||
| AK016859.1 | - | 7 | 744 | |||
| AK077182.1 | - | 3 | 618 | |||
| BC107290.1 | - | 2 | 146 | 40053043 | ||
| BC107291.1 | - | 2 | 146 | 40053045 | ||

Lrrc72
Genomic Map