siRNA Details
| Gene Symbol: 4933421I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027702.2 | NP_081978.1 | 2 | 233 | ||
| GenBank mRNA | AK016864.1 | - | 2 | 234 | ||
| BC147587.1 | - | 2 | 273 | 9056577 | ||
| BC147628.1 | - | 2 | 273 | 9056618 | ||

4933421I07Rik
Genomic Map