siRNA Details
| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138757.2 | NP_620096.2 | 4 | 949 | ||
| GenBank mRNA | BC026851.1 | - | 4 | 878 | 4459471 | |
| BC027229.1 | - | 4 | 534 | 2615769 | ||
| CD560819.1 | - | 2 | 156 | |||
| CF167981.1 | - | 4 | 545 | |||

Asun
Genomic Map