siRNA Details
| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138757.2 | NP_620096.2 | 4 | 1029 | ||
| GenBank mRNA | BC026851.1 | - | 4 | 958 | 4459471 | |
| BC027229.1 | - | 4 | 614 | 2615769 | ||
| CD560819.1 | - | 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asun| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138757.2 | NP_620096.2 | 4 | 1029 | ||
| GenBank mRNA | BC026851.1 | - | 4 | 958 | 4459471 | |
| BC027229.1 | - | 4 | 614 | 2615769 | ||
| CD560819.1 | - | 2 | 236 | |||