siRNA Details
| Gene Symbol: Otud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027715.1 | NP_081991.1 | 1 | 1085 | ||
| GenBank mRNA | AK016972.1 | - | 1 | 392 | ||
| AK206709.1 | - | 1 | 39 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud1| Gene Symbol: Otud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027715.1 | NP_081991.1 | 1 | 1085 | ||
| GenBank mRNA | AK016972.1 | - | 1 | 392 | ||
| AK206709.1 | - | 1 | 39 | |||