siRNA Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027725.3 | NP_082001.2 | 8 | 708 | ||
| GenBank mRNA | AK016201.1 | - | 11 | 998 | ||
| AK016977.1 | - | 8 | 709 | |||
| BC049704.1 | - | 8 | 708 | |||
| BC144994.1 | - | 8 | 685 | |||
| BC145995.1 | - | 8 | 708 | 40131298 | ||

Daw1
Genomic Map