siRNA Details
| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027733.5 | NP_082009.3 | 2 | 201 | ||
| NM_029485.2 | NP_083761.1 | 2 | 201 | |||
| GenBank mRNA | AK016341.1 | - | 2 | 207 | ||
| AY730532.1 | - | 2 | 201 | |||
| BC060950.1 | - | 2 | 154 | 6773733 | ||
| BU961109.1 | - | 2 | 204 | |||

Spata24
Genomic Map