siRNA Details
| Gene Symbol: Spata24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027733.5 | NP_082009.3 | 1, 2 | 176 | ||
| NM_029485.2 | NP_083761.1 | 1, 2 | 176 | |||
| GenBank mRNA | AK017218.1 | - | 1, 2 | 152 | ||
| AY730532.1 | - | 1, 2 | 176 | |||
| BC048524.1 | - | 1, 2 | 125 | |||
| BC060950.1 | - | 1, 2 | 129 | 6773733 | ||
| BU961109.1 | - | 1, 2 | 179 | |||

Spata24
Genomic Map