siRNA Details
| Gene Symbol: Mro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017067.1 | - | 3, 4 | 341 | ||
| AY243876.1 | - | 6, 7 | 751 | |||
| CK335185.1 | - | 5, 6 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mro| Gene Symbol: Mro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017067.1 | - | 3, 4 | 341 | ||
| AY243876.1 | - | 6, 7 | 751 | |||
| CK335185.1 | - | 5, 6 | 614 | |||