siRNA Details
| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027745.1 | NP_082021.1 | 3 | 636 | ||
| GenBank mRNA | AK220306.1 | - | 3 | 619 | ||
| BC056480.1 | - | 3 | 636 | 5689542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc57| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027745.1 | NP_082021.1 | 3 | 636 | ||
| GenBank mRNA | AK220306.1 | - | 3 | 619 | ||
| BC056480.1 | - | 3 | 636 | 5689542 | ||