siRNA Details
Gene Symbol: Ccdc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027745.1 | NP_082021.1 | 9 | 1485 | ||
GenBank mRNA | BC056480.1 | - | 9 | 1485 | 5689542 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027745.1 | NP_082021.1 | 9 | 1485 | ||
GenBank mRNA | BC056480.1 | - | 9 | 1485 | 5689542 |