siRNA Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002438.3 | NP_002429.1 | 2 | 525 | ||
| GenBank mRNA | AK308132.1 | - | 2 | 526 | ||
| AK308167.1 | - | 2 | 521 | |||
| BC142642.1 | - | 2 | 506 | |||
| BC146838.1 | - | 2 | 448 | 9056765 | ||
| DC296466.1 | - | 2 | 525 | |||
| DQ663787.1 | - | 2 | 402 | |||
| J05550.1 | - | 2 | 505 | |||
| X55635.1 | - | 2 | 478 | |||

MRC1
Genomic Map