siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017076.1 | - | 1 | 415 | ||
| AK181465.1 | - | 39 | ||||
| CX205694.1 | - | 1 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crnde| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017076.1 | - | 1 | 415 | ||
| AK181465.1 | - | 39 | ||||
| CX205694.1 | - | 1 | 125 | |||