siRNA Details
| Gene Symbol: Rassf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027760.2 | NP_082036.1 | 1, 2 | 181 | ||
| GenBank mRNA | AF424737.1 | - | 1, 2 | 181 | ||
| BC115779.1 | - | 1, 2 | 152 | 40105476 | ||
| BC117546.1 | - | 1, 2 | 152 | 40105479 | ||

Rassf8
Genomic Map