siRNA Details
| Gene Symbol: Tprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175165.3 | NP_780374.1 | 6 | 969 | ||
| GenBank mRNA | AK036796.1 | - | 6 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tprg| Gene Symbol: Tprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175165.3 | NP_780374.1 | 6 | 969 | ||
| GenBank mRNA | AK036796.1 | - | 6 | 972 | ||