siRNA Details
| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024242.3 | NP_077204.2 | 9 | 1273 | ||
| GenBank mRNA | AK017312.1 | - | 9 | 968 | ||
| AK152707.1 | - | 9 | 1022 | |||
| AK161315.1 | - | 10 | 1516 | |||
| BC002158.1 | - | 3 | 230 | 3485522 | ||

Riok1
Genomic Map