siRNA Details
| Gene Symbol: Ano9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178381.3 | NP_848468.2 | 9 | 832 | ||
| GenBank mRNA | BC025477.1 | - | 9 | 800 | ||
| BC049815.1 | - | 2 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano9| Gene Symbol: Ano9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178381.3 | NP_848468.2 | 9 | 832 | ||
| GenBank mRNA | BC025477.1 | - | 9 | 800 | ||
| BC049815.1 | - | 2 | 510 | |||