siRNA Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290521.1 | NP_001277450.1 | 5 | 594 | ||
| NM_001290522.1 | NP_001277451.1 | 5 | 484 | |||
| NM_023597.2 | NP_076086.1 | 5 | 487 | |||
| GenBank mRNA | AF274321.1 | - | 5 | 464 | ||
| AK017259.1 | - | 5 | 477 | |||
| AK141026.1 | - | 5 | 592 | |||
| BC026500.1 | - | 4 | 373 | |||
| BC037102.1 | - | 5 | 477 | 5364307 | ||
| CK618879.1 | - | 5 | 594 | |||

Wdr31
Genomic Map