siRNA Details
| Gene Symbol: Col24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027770.2 | NP_082046.2 | 3 | 1022 | ||
| GenBank mRNA | AY243578.1 | - | 1 | 176 | ||
| BC139134.1 | - | 3 | 579 | 8862156 | ||
| BC145544.1 | - | 3 | 579 | |||
| DQ157748.1 | - | 3 | 1022 | |||

Col24a1
Genomic Map