siRNA Details
| Gene Symbol: Col24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027770.2 | NP_082046.2 | 17, 18 | 2903 | ||
| GenBank mRNA | BC139134.1 | - | 17, 18 | 2460 | 8862156 | |
| BC145544.1 | - | 17, 18 | 2452 | |||
| DQ157748.1 | - | 17, 18 | 2903 | |||

Col24a1
Genomic Map