siRNA Details
Gene Symbol: Chst9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199055.2 | NP_951010.1 | 4 | 758 | ||
GenBank mRNA | AB106879.1 | - | 4 | 758 | ||
AK017407.1 | - | 6 | 1071 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chst9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199055.2 | NP_951010.1 | 4 | 758 | ||
GenBank mRNA | AB106879.1 | - | 4 | 758 | ||
AK017407.1 | - | 6 | 1071 |