siRNA Details
| Gene Symbol: Chst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199055.2 | NP_951010.1 | 4 | 1103 | ||
| GenBank mRNA | AB106879.1 | - | 4 | 1103 | ||
| AK017407.1 | - | 6 | 1416 | |||
| BC107361.1 | - | 4 | 1101 | 40056052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst9| Gene Symbol: Chst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199055.2 | NP_951010.1 | 4 | 1103 | ||
| GenBank mRNA | AB106879.1 | - | 4 | 1103 | ||
| AK017407.1 | - | 6 | 1416 | |||
| BC107361.1 | - | 4 | 1101 | 40056052 | ||