siRNA Details
| Gene Symbol: Pex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293806.1 | NP_001280735.1 | 5 | 986 | ||
| NM_027777.2 | NP_082053.1 | 5 | 986 | |||
| GenBank mRNA | AK149599.1 | - | 5 | 688 | ||
| BC090845.1 | - | 5 | 986 | 6400629 | ||

Pex1
Genomic Map