siRNA Details
| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 21 | 2841 | ||
| GenBank mRNA | AF513716.1 | - | 10 | 1238 | ||
| AK080318.1 | - | 1 | 16 | |||
| AK173293.1 | - | 4 | 188 | |||
| BC048004.1 | - | 6 | 729 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmnl2| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 21 | 2841 | ||
| GenBank mRNA | AF513716.1 | - | 10 | 1238 | ||
| AK080318.1 | - | 1 | 16 | |||
| AK173293.1 | - | 4 | 188 | |||
| BC048004.1 | - | 6 | 729 | |||