siRNA Details
| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 12 | 1365 | ||
| GenBank mRNA | CA329111.1 | - | 8 | 736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmnl2| Gene Symbol: Fmnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172409.2 | NP_765997.2 | 12 | 1365 | ||
| GenBank mRNA | CA329111.1 | - | 8 | 736 | ||