siRNA Details
| Gene Symbol: Amtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027793.1 | NP_082069.1 | 8 | 531 | ||
| GenBank mRNA | AK017352.1 | - | 8 | 531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amtn| Gene Symbol: Amtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027793.1 | NP_082069.1 | 8 | 531 | ||
| GenBank mRNA | AK017352.1 | - | 8 | 531 | ||