siRNA Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144877.1 | NP_659126.1 | 6, 7 | 1790 | ||
| GenBank mRNA | AK084750.1 | - | 6, 7 | 1485 | ||
| AK146751.1 | - | 1988 | ||||
| AK168959.1 | - | 6, 7 | 1997 | |||
| AK169319.1 | - | 1989 | ||||
| AK173044.1 | - | 1943 | ||||
| BC014872.1 | - | 6, 7 | 1790 | 3602469 | ||

Mettl13
Genomic Map