siRNA Details
| Gene Symbol: Obox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027802.2 | NP_082078.1 | 4 | 574 | ||
| GenBank mRNA | AK018362.1 | - | 4 | 571 | ||
| AK136061.1 | - | 4 | 564 | |||
| BC141323.1 | - | 534 | 9055935 | |||
| BC141324.1 | - | 534 | 9055936 | |||

Obox1
Genomic Map